Cerium(III)-2-ethylhexanoate 49% (12% Ce) in 2-ethylhexanoic acid

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cerium(III) 2-ethylhexanoate

40451-22 40451-36 40451-A3
CAAA40451-22EA 40.45 CAD
CAAA40451-22 CAAA40451-36 CAAA40451-A3
Cerium(III)-2-ethylhexanoate 49% (12% Ce) in 2-ethylhexanoic acid
Cerium (III) 2-ethylhexanoate

CAS No.: 56797-01-4 Molecular Formula: C24H45CeO6 Formula Weight: 569.74 Flash Point: 110°(230°F) Physical Form: Viscous Liquid Density: 1.08 MDL No.: MFCD00014004

Formula: C₂₄H₄₅CeO₆
MW: 569.727 g/mol
Density: 1.08
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014004
CAS Number: 56797-01-4
EINECS: 260-386-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR