Potassium hexachlororuthenate(IV) (metal basis) (≥25,3% Ru) ≥99.95%, Premion®
Supplier: Thermo Scientific Chemicals
|
Warning
|
40399-03
40399-06
CAAA40399-03EA
259.12
CAD
CAAA40399-03
CAAA40399-06
Potassium hexachlororuthenate(IV) (metal basis) (≥25,3% Ru) ≥99.95%, Premion®
Potassium hexachlororuthenate(IV) (metal basis)
Purity: 99.95%
CAS No.: 23013-82-3
Molecular Formula: K2RuCl6
Formula Weight: 391.99
Physical Form: Cystalline Powder
MDL No.: MFCD00050159
Formula:
K₂RuCl₆ MW: 391.99 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00050159 CAS Number: 23013-82-3 EINECS: 245-381-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...