Cobalt naphthenate ∼53% (6% Co) in mineral spirits
Supplier: Thermo Scientific Chemicals
|
Danger
|
40387-36
40387-A3
CAAA40387-36EA
251.22
CAD
CAAA40387-36
CAAA40387-A3
Cobalt naphthenate ∼53% (6% Co) in mineral spirits
Cobalt naphthenate
CAS No.: 61789-51-3
Flash Point: 40°(104°F)
Physical Form: Dark Liquid
UN No.: 1993
MDL No.: MFCD02094763
Formula:
C₂₂H₁₄CoO₄ Flash Pt: 40 °C (104 °F) Storage Temperature: Ambient |
MDL Number:
MFCD02094763 CAS Number: 61789-51-3 EINECS: 263-064-0 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...