Silica gel, granules ACS -4+10 Mesh desiccant, with indicator

Supplier: Thermo Scientific Chemicals
Danger

40381-22 40381-36 40381-A4
CAAA40381-22EA 71.42 CAD
CAAA40381-22 CAAA40381-36 CAAA40381-A4
Silica gel, granules ACS -4+10 Mesh desiccant, with indicator
Silica gel

CAS No.: 7631-86-9 Physical Form: -4+10 Mesh Granules Sensitivity: Hygroscopic pH: 6.0 MDL No.: MFCD00011232

Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230
Melting Pt: 1710
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
EINECS: 231-545-4
Merck Index: 14,8490

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR