Aluminum, silver coated (19 - 21% Ag) (w/w) ≥99.9% (metals basis), powder -200 mesh
Supplier: Thermo Scientific Chemicals
40340-14
40340-22
CAAA40340-14EA
109.81
CAD
CAAA40340-14
CAAA40340-22
Aluminum, silver coated (19 - 21% Ag) (w/w) ≥99.9% (metals basis), powder -200 mesh
Aluminum, silver coated
Applications: Conductive adhesives
Purity: 99.9%
Physical Form: Density 1.1-1.7g/cm3
Odor: Odorless
MDL No.: MFCD05865335
Applications: Conductive adhesives
Storage Temperature:
Ambient |
MDL Number:
MFCD05865335 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...