Polyethyleneimine, M.W. 10,000 ≥99%, branched
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Polyaziridine, Corcat, Polyethylenimine, Polymin P, Polyaziridine Mw 25000, Aziridine, PEI, Polyethylenimine (branched), Ethyleneimine Polymer, Poly(ethyleneimine)
40331-14
40331-22
40331-36
CAAA40331-14EA
82.67
CAD
CAAA40331-14
CAAA40331-22
CAAA40331-36
Polyethyleneimine, M.W. 10,000 ≥99%, branched
Polyethyleneimine
Purity: 99%
CAS No.: 9002-98-6
Molecular Formula: (-NHCH2CH2-)x(-N(CH2CH2NH2)CH2CH2-)y
Physical Form: Liquid
Appearance: Pale yellow
Odor: Amine-like
MDL No.: MFCD00803910
Formula:
(CH2CH2NH)n Storage Temperature: Ambient |
MDL Number:
MFCD00084427 CAS Number: 9002-98-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...