Potassium fluorosulfate ≥99.5%
Supplier: Thermo Scientific Chemicals
|
Danger
|
40322-09
40322-18
CAAA40322-09EA
241.74
CAD
CAAA40322-09
CAAA40322-18
Potassium fluorosulfate ≥99.5%
Potassium fluorosulfate
Purity: 99.5%
CAS No.: 13455-22-6
Molecular Formula: FKSO3
Formula Weight: 138.16
Physical Form: Powder/lump
Sensitivity: Moisture Sensitive
Appearance: White
Melting Point: 350° dec.
UN No.: 3260
MDL No.: MFCD00216618
Formula:
FKSO₃ Melting Pt: 350 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00216618 CAS Number: 13455-22-6 UN: 3260 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...