Sodium ammonium hydrogen phosphate tetrahydrate, Reagent Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
XSodium ammonium hydrogen phosphate tetrahydrate, Sodium ammonium phosphate, Ammonium sodium phosphate dibasic tetrahydrate, Sodium ammonium phosphate tetrahydrate, Ammonium sodium phosphate tetrahydrate
Soluble in water. Insoluble in alcohol
CAS No.: 7783-13-3
Molecular Formula: NaNH4HPO4.4H2O
Formula Weight: 209.07 (137.02anhy)
Physical Form: Crystalline
Density: 1.544
Odor: Odorless
Melting Point: 79° dec.
Solubility: Soluble in water. Insoluble in alcohol
MDL No.: MFCD00066970
Formula:
NaNH₄HPO₄·4H₂O MW: 209.07 g/mol Melting Pt: 79 °C (dec.) Density: 1.544 Storage Temperature: Ambient |
MDL Number:
MFCD00066970 CAS Number: 7783-13-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...