Neodymium (III) sulfate octahydrate ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
40253-14
40253-22
CAAA40253-14EA
122.77
CAD
CAAA40253-14
CAAA40253-22
Neodymium (III) sulfate octahydrate ≥99.9% (REO, rare earth oxide basis)
Neodymium (III) sulphate octahydrate
Soluble in water
Purity: 99.9%
CAS No.: 13477-91-3
Molecular Formula: Nd2(SO4)3.8H2O
Formula Weight: 720.78 (576.66anhy)
Physical Form: Crystalline
Density: 2.85
Melting Point: 1176°
Solubility: Soluble in water
Merck Reference: 14,6450
MDL No.: MFCD00149803
Formula:
Nd₂S₃O₁₂·8H₂O MW: 720.78 g/mol (576.66 anhy) Melting Pt: 1176 °C Density: 2.85 Storage Temperature: Ambient |
MDL Number:
MFCD00149803 CAS Number: 13477-91-3 EINECS: 233-262-1 Merck Index: 14,6450 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...