Strontium molybdenum oxide 99%
Supplier: Thermo Scientific Chemicals
40238-09
40238-18
40238-30
40238-A1
CAAA40238-09EA
38.55
CAD
CAAA40238-09
CAAA40238-18
CAAA40238-30
CAAA40238-A1
Strontium molybdenum oxide 99%
Strontium molybdenum oxide
Insoluble in water. Soluble in acid
Purity: 99.9%
CAS No.: 13470-04-7
Molecular Formula: SrMoO4
Formula Weight: 247.56
Physical Form: -200 Mesh Powder
Density: 4.540
Melting Point: dec.
Solubility: Insoluble in water. Soluble in acid
MDL No.: MFCD00049550
Formula:
SrMoO₄ Melting Pt: Dec. Density: 4,540 Storage Temperature: Ambient |
MDL Number:
MFCD00049550 CAS Number: 13470-04-7 EINECS: 236-730-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...