Zinc(II) wolframate ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Zinc tungsten oxide
40234-09
40234-18
CAAA40234-09EA
119.92
CAD
CAAA40234-09
CAAA40234-18
CAAA40234-30
Zinc(II) wolframate ≥99.9% (metals basis)
Zinc(II) wolframate
Purity: 99.9%
CAS No.: 13597-56-3
Molecular Formula: ZnWO4
Formula Weight: 313.22
Physical Form: -200 Mesh Powder
Formula:
O₄WZn MW: 313.22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00049633 CAS Number: 13597-56-3 EINECS: 237-053-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...