Cerium(III) oxalate hydrate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cerium(III) oxalate hydrate

40227-14 40227-22
CAAA40227-14EA 117.71 CAD
CAAA40227-14 CAAA40227-22
Cerium(III) oxalate hydrate 99%
Cerium (III) oxalate hydrate

Insoluble in water. Soluble in hot dilute HCl and H2SO3


Purity: 99% CAS No.: 15750-47-7 Molecular Formula: Ce2(C2O4)3.xH2O Formula Weight: 544.29(anhy) Physical Form: Crystalline powder Sensitivity: Hygroscopic Solubility: Insoluble in water. Soluble in hot dilute HCl and H2SO4 Merck Reference: 14,2001 UN No.: 3288 MDL No.: MFCD00150139

Formula: C₆Ce₂O₁₂·H₂O
MW: 544.29 g/mol (anhy)
Melting Pt: Dec.
Storage Temperature: Ambient
MDL Number: MFCD00150139
CAS Number: 15750-47-7
UN: 3288
ADR: 6.1,III
Merck Index: 14,2001

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR