Cobalt(II) wolframate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
40223-09
40223-18
40223-30
CAAA40223-09EA
79.16
CAD
CAAA40223-09
CAAA40223-18
CAAA40223-30
Cobalt(II) wolframate 99%
Cobalt(II) wolframate
Applications: pigment, drier for enamels, inks, and paints, electronic devices
Insoluble in water. Soluble in hot concentrated acids
Purity: 99%
CAS No.: 10101-58-3
Molecular Formula: CoWO4
Formula Weight: 306.78
Physical Form: Powder
Density: 8.42
Solubility: Insoluble in water. Soluble in hot concentrated acids
MDL No.: MFCD00016034
pigment, drier for enamels, inks, and paints, electronic devices
Formula:
CoO₄W Density: 8,42 Storage Temperature: Ambient |
MDL Number:
MFCD00016034 CAS Number: 10101-58-3 EINECS: 233-254-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...