Copper(II) wolframate ≥99.5% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
40217-18
40217-30
CAAA40217-18EA
200.66
CAD
CAAA40217-18
CAAA40217-30
Copper(II) wolframate ≥99.5% (metals basis)
Copper(II) wolframate
Purity: 99.5%
CAS No.: 13587-35-4
Molecular Formula: CuWO4
Formula Weight: 311.38
Physical Form: -200 Mesh Powder
Odor: Odorless
Merck Reference: 14,2657
MDL No.: MFCD00016067
Formula:
CuO₄W Storage Temperature: Ambient |
MDL Number:
MFCD00016067 CAS Number: 13587-35-4 EINECS: 237-022-7 Merck Index: 14,2657 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...