Hexachloroplatinic(IV) acid solution (20% Pt)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hydrogen hexachloroplatinate (IV), Dihydrogen hexachloroplatinate (IV)

40177-NA 40177-NJ
CAAA40177-NAEA 1139.18 CAD
CAAA40177-NA CAAA40177-NJ
Hexachloroplatinic(IV) acid solution (20% Pt)
Hexachloroplatinic (IV) acid

CAS No.: 16941-12-1 Molecular Formula: H2Cl6Pt Formula Weight: 409.82(anhy) Physical Form: Liquid Appearance: Red-brown UN No.: 3264 MDL No.: MFCD00011330

Formula: H₂Cl₆Pt
Storage Temperature: Ambient
MDL Number: MFCD00011330
CAS Number: 16941-12-1
EINECS: 241-010-7
UN: 3264
ADR: 8,III
Merck Index: 14,7526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR