Calcium iodate monohydrate ≥99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
40138-22
40138-36
CAAA40138-22EA
82.63
CAD
CAAA40138-22
CAAA40138-36
Calcium iodate monohydrate ≥99%
Calcium iodate monohydrate
Slightly soluble in water
Purity: 99+%
CAS No.: 10031-32-0
Molecular Formula: Ca(IO3)2.H2O
Formula Weight: 407.90 (389.88anhy)
Physical Form: Powder
Solubility: Slightly soluble in water
UN No.: 1479
MDL No.: MFCD00036265
Formula:
Ca(IO₃)₂·H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00036265 CAS Number: 10031-32-0 EINECS: 232-191-3 UN: 1479 ADR: 5.1,II Merck Index: 14,1676 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...