Zirconium(IV) oxide 18% in water stabilized, colloidal dispersion

Supplier: Thermo Scientific Chemicals
Danger


Stabilization: Stabilised with yttria (Y₂O₃) 1.3%

40123-36 40123-A4
CAAA40123-36EA 208.56 CAD
CAAA40123-36 CAAA40123-A4
Zirconium(IV) oxide 18% in water stabilized, colloidal dispersion
Zirconium (IV) oxide

Counter ion mol/mol 0.3 nitrate, pH 5-6, particle charge (+), viscosity approx. 20 cps


Applications: In ceramic industry


Applications: In ceramic industry

Formula: O₂Zr
Storage Temperature: Ambient
MDL Number: MFCD00011310
CAS Number: 1314-23-4
EINECS: 215-227-2
Merck Index: 14,10180

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR