Indium(III) hydroxide ≥99.8% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
40100-09
40100-18
CAAA40100-09EA
117.39
CAD
CAAA40100-09
CAAA40100-18
Indium(III) hydroxide ≥99.8% (metals basis)
Indium (III) hydroxide
Purity: 99.8%
CAS No.: 20661-21-6
Molecular Formula: In(OH)3
Formula Weight: 165.84
Physical Form: Powder
Melting Point: 150° -H2O
MDL No.: MFCD00011056
Formula:
H₃InO₃ Melting Pt: 150 °C (-H₂O) Storage Temperature: Ambient |
MDL Number:
MFCD00011056 CAS Number: 20661-21-6 EINECS: 243-947-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...