Tantalum(V) isopropoxide 10% (w/v) in 2-propanol/hexane , 99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

40038-AC 40038-AE
CAAA40038-ACEA 225.94 CAD
CAAA40038-AC CAAA40038-AE CAAA40038-AP
Tantalum(V) isopropoxide 10% (w/v) in 2-propanol/hexane , 99.9% (metals basis)
Tantalum (V) isopropoxide

Purity: 99.9% CAS No.: 16761-83-4 Molecular Formula: C15H35O5Ta Formula Weight: 476.39 Physical Form: Liquid Sensitivity: Moisture Sensitive UN No.: 1993 MDL No.: MFCD00145347

Formula: C₁₅H₃₅O₅Ta
Storage Temperature: Ambient
MDL Number: MFCD00145347
CAS Number: 16761-83-4
UN: 1993
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR