Palladium 5% on strontium carbonate, powder, reduced

Supplier: Thermo Scientific Chemicals

39819-04 39819-09 39819-18
CAAA39819-04EA 54.19 CAD
CAAA39819-04 CAAA39819-09 CAAA39819-18
Palladium 5% on strontium carbonate, powder, reduced
Palladium
Formula: Pd
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR