Lead zirconium titanium oxide (∼30 wt% Oxide) polymeric precursor

Supplier: Thermo Scientific Chemicals
Danger

39758-09 39758-18
CAAA39758-09EA 149.78 CAD
CAAA39758-09 CAAA39758-18 CAAA39758-30
Lead zirconium titanium oxide (∼30 wt% Oxide) polymeric precursor
Lead zirconium titanium oxide

Applications: piezoelectric devices and electro-optic devices
Soluble in alcohols


CAS No.: 12626-81-2 Physical Form: Semi-solid/viscous liquid Appearance: Pale yellow Solubility: Soluble in alcohols UN No.: 1993 MDL No.: MFCD00212270


Used in piezoelectric devices and electro-optic devices

Formula: PbZr(1-x)TixO₃
Storage Temperature: Ambient
MDL Number: MFCD00212270
CAS Number: 12626-81-2
EINECS: 235-727-4
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR