Sodium aurothiomalate ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

39740-03 39740-06
CAAA39740-03EA 605.14 CAD
CAAA39740-03 CAAA39740-06
Sodium aurothiomalate ≥99.9% (metals basis)
Sodium aurothiomalate

Very soluble in water. Practically insoluble in alcohol, ether


Purity: 99.9% CAS No.: 12244-57-4 Molecular Formula: C4H3AuNa2O4S Formula Weight: 390.08 Physical Form: Powder Odor: Sulphurous Merck Reference: 14,4518 MDL No.: MFCD00064304

Formula: C₄H₃AuNa₂O₄S
Storage Temperature: Ambient
MDL Number: MFCD00064304
CAS Number: 12244-57-4
EINECS: 235-479-7
Merck Index: 14,4518

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR