Dihydrogen hexafluorotitanate 60% (w/w) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

39736-36 39736-A1
CAAA39736-36EA 141.57 CAD
CAAA39736-36 CAAA39736-A1
Dihydrogen hexafluorotitanate 60% (w/w) in aqueous solution
Dihydrogen hexafluorotitanate

CAS No.: 17439-11-1 Molecular Formula: H2TiF6 Formula Weight: 163.91 Physical Form: Liquid Density: 1.69 Odor: Pungent UN No.: 2922 MDL No.: MFCD00049647

Formula: H2TiF6
Density: 1.69
Storage Temperature: Ambient
MDL Number: MFCD00049647
CAS Number: 17439-11-1
EINECS: 241-460-4
UN: 2922
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR