Carbon black ≥99.9%, acetylene, 50% compressed

Supplier: Thermo Scientific Chemicals

Synonyms: Furnace black, Lamp Black

39724-30 39724-A1 39724-A5 039724-18
CAAA39724-30EA 108.07 CAD
CAAA39724-30 CAAA39724-A1 CAAA39724-A5 AA39724-18
Carbon black ≥99.9%, acetylene, 50% compressed
Carbon black

Average particle size 0.042 micron


Purity: 99.9+% CAS No.: 1333-86-4 Physical Form: S.A. 75m2/g, Bulk density 80-120g/L Appearance: Black Odor: Odorless UN No.: 1325 MDL No.: MFCD00133992

Formula: C
MW: 12.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 1333-86-4
EINECS: 215-609-9
UN: 1325
ADR: 4.1,III

Order Now

Specification Test Results

Trace Metal 99.9+%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR