Titanium(III) sulfate 20% in 1-4% sulphuric acid

Supplier: Thermo Scientific Chemicals
Danger

39664-30 39664-A1 39664-A7
CAAA39664-30EA 116.45 CAD
CAAA39664-30 CAAA39664-A1 CAAA39664-A7
Titanium(III) sulfate 20% in 1-4% sulphuric acid
Titanium(III) sulfate

CAS No.: 10343-61-0 Molecular Formula: Ti2(SO4)3 Formula Weight: 383.38 Physical Form: Liquid Density: 1.15 UN No.: 3264 MDL No.: MFCD00036317

Formula: O₁₂S₃Ti₂
Density: 1.15
Storage Temperature: Ambient
MDL Number: MFCD00036317
CAS Number: 10343-61-0
UN: 3264
ADR: 8,III
Merck Index: 14,9476

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR