Sodium hexacyanoferrate(II) decahydrate ≥99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Sodium ferrocyanide decahydrate
39660-18
39660-30
CAAA39660-18EA
110.13
CAD
CAAA39660-18
CAAA39660-30
Sodium hexacyanoferrate(II) decahydrate ≥99%
Sodium hexacyanoferrate(II) decahydrate
Soluble in water
Purity: 99%
CAS No.: 14434-22-1
Molecular Formula: Na4Fe(CN)6.10H2O
Formula Weight: 484.07 (303.92anhy)
Physical Form: Powder
Density: 1.458
Odor: Odorless
Melting Point: 82° -10H2O
Solubility: Soluble in water
MDL No.: MFCD00085360
Formula:
C₆H₂₀FeN₆Na₄O₁₀ Melting Pt: 82 °C (-10H₂O) Density: 1.458 Storage Temperature: Ambient |
MDL Number:
MFCD00085360 CAS Number: 14434-22-1 EINECS: 237-081-9 Merck Index: 14,8616 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...