Silver molybdenum oxide ≥99%
Supplier: Thermo Scientific Chemicals
39650-01
39650-03
CAAA39650-01EA
90.06
CAD
CAAA39650-01
CAAA39650-03
Silver molybdenum oxide ≥99%
Silver molybdenum oxide
Purity: 99%
CAS No.: 13765-74-7
Molecular Formula: Ag2MoO4
Formula Weight: 375.68
Physical Form: Powder
Density: 6.18
Sensitivity: Light Sensitive
Appearance: Beige
Odor: Odorless
Melting Point: 483°
MDL No.: MFCD00053384
Formula:
Ag₂MoO₄ MW: 375.68 g/mol Melting Pt: 483 °C Density: 6,18 Storage Temperature: Ambient |
MDL Number:
MFCD00053384 CAS Number: 13765-74-7 EINECS: 237-374-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...