Nickel(II) citrate hydrate ≥98%

Supplier: Thermo Scientific Chemicals
Danger

39634-22 39634-36
CAAA39634-22EA 131.14 CAD
CAAA39634-22 CAAA39634-36
Nickel(II) citrate hydrate ≥98%
Nickel(II) citrate hydrate

Purity: 98+% CAS No.: 6018-92-4 Molecular Formula: Ni3(C6H5O7)2.xH2O Formula Weight: 554.33(anhy) Physical Form: Powder UN No.: 3077 MDL No.: MFCD00054366

Formula: Ni₃(C₆H₅O₇)₂.xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00054366
CAS Number: 6018-92-4
EINECS: 227-873-2
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR