(Pentamethylcyclopentadienyl)titanium trichloride

Supplier: Thermo Scientific Chemicals
Danger

39596-03 39596-06
CAAA39596-03EA 178.54 CAD
CAAA39596-03 CAAA39596-06
(Pentamethylcyclopentadienyl)titanium trichloride
(Pentamethylcyclopentadienyl)titanium trichloride

CAS No.: 12129-06-5 Molecular Formula: C10H15Cl3Ti Formula Weight: 289.47 Physical Form: Crystalline Sensitivity: Moisture Sensitive Appearance: Red Melting Point: 200° dec. UN No.: 3261 MDL No.: MFCD00070434

Formula: C₁₀H₁₅Cl₃Ti
Melting Pt: 200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00070434
CAS Number: 12129-06-5
UN: 3261
ADR: 8,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR