Titanium(IV) oxide sulfate sulfuric acid hydrate

Supplier: Thermo Scientific Chemicals
Danger

39591-36 39591-A3
CAAA39591-36EA 186.44 CAD
CAAA39591-36 CAAA39591-A3
Titanium(IV) oxide sulfate sulfuric acid hydrate

CAS No.: 13825-74-6 Molecular Formula: TiOSO4.xH2O + H2SO4 Physical Form: -10 Mesh Powder UN No.: 3260 MDL No.: MFCD00011270

Formula: TiOSO₄·xH₂SO₄·yH₂O
Storage Temperature: Ambient
MDL Number: MFCD00150565
CAS Number: 123334-00-9
EINECS: 237-523-0
UN: 3260
ADR: 8,II
Merck Index: 14,9475

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR