Vanadium(IV) oxide phthalocyanine

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Vanadyl phthalocyanine

39587-02 39587-04 39587-09
CAAA39587-02EA 62.57 CAD
CAAA39587-02 CAAA39587-04 CAAA39587-09
Vanadium(IV) oxide phthalocyanine
Vanadium(IV) oxide phthalocyanine

CAS No.: 13930-88-6 Molecular Formula: C32H16N8OV Formula Weight: 579.45 Physical Form: Crystalline Appearance: Dark blue to purple UN No.: 3285 MDL No.: MFCD00041966

Formula: C₃₂H₁₆N₈OV
MW: 579.45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00041966
CAS Number: 13930-88-6
EINECS: 237-700-2
UN: 3285
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR