Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato-O,O')ytterbium(III) ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
Synonyms:
Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionate)ytterbium(III), Yb(FOD)3, Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)ytterbium(III)
39582-03
39582-06
CAAA39582-03EA
142.83
CAD
CAAA39582-03
CAAA39582-06
Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato-O,O')ytterbium(III) ≥99% (metals basis)
Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato-O,O')ytterbium(III)
Purity: 99%
CAS No.: 18323-96-1
Molecular Formula: C30H30F21O6Yb
Formula Weight: 1058.59
Sensitivity: Hygroscopic
Melting Point: 108-111°
MDL No.: MFCD00044289
Formula:
C₃₀H₃₀F₂₁O₆Yb MW: 1058.562 g/mol Melting Pt: 108…111 °C Storage Temperature: Ambient |
MDL Number:
MFCD00044289 CAS Number: 18323-96-1 EINECS: 242-211-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...