Ytterbium(III) isopropoxide
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ytterbium(III) i-propoxide
39579-03
39579-06
CAAA39579-03EA
152
CAD
CAAA39579-03
CAAA39579-06
Ytterbium(III) isopropoxide
Ytterbium (III) isopropoxide
CAS No.: 6742-69-4
Molecular Formula: C9H21O3Yb
Formula Weight: 350.31
Physical Form: Powder, Ampouled under argon
Sensitivity: Air and Moisture Sensitive
Odor: Odorless
Boiling Point: 190-200°/0.2mm subl.
UN No.: 2925
MDL No.: MFCD00145532
Formula:
C₉H₂₁O₃Yb MW: 350.30 g/mol Boiling Pt: 190…200 °C (0,2 mmHg) (subl.) Melting Pt: 200 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00145532 CAS Number: 6742-69-4 UN: 2925 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...