Dicarbonylbis(cyclopentadienyl)titanium(II) ≥98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Bis(cyclopentadienyl)dicarbonyl titanium(II)

39571-02 39571-04
CAAA39571-02EA 183.28 CAD
CAAA39571-02 CAAA39571-04
Dicarbonylbis(cyclopentadienyl)titanium(II) ≥98%
Dicarbonylbis(cyclopentadienyl)titanium(II)

Purity: 98% CAS No.: 12129-51-0 Molecular Formula: C12H10O2Ti Formula Weight: 234.09 Physical Form: Crystalline Sensitivity: Air and Moisture Sensitive Melting Point: 90° dec. Boiling Point: 60°/0.001mm subl. UN No.: 3466 MDL No.: MFCD00143762

Formula: C₁₂H₁₀O₂Ti
MW: 234.076 g/mol
Boiling Pt: 60 °C (0.001 mmHg) (subl.)
Melting Pt: 90 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00143762
CAS Number: 12129-51-0
UN: 3466
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR