Zinc 2-ethylhexanoate ∼80% (17 - 19% Zn) in mineral spirits

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Zinc(II) 2-ethylhexanoate

39561-36 39561-0E 39561-22
CAAAA39561-36EA 125.93 CAD
CAAAA39561-36 CAAA39561-0E CAAA39561-22
Zinc 2-ethylhexanoate ∼80% (17 - 19% Zn) in mineral spirits
Zinc 2-ethylhexanoate

CAS No.: 136-53-8 Molecular Formula: C16H30O4Zn Formula Weight: 351.79 Physical Form: Liquid Density: 1.07 Appearance: Colorless UN No.: 1268 MDL No.: MFCD00053316

Formula: C₁₆H₃₀O₄Zn
MW: 351.792 g/mol
Density: 1.07
Storage Temperature: Ambient
MDL Number: MFCD00053316
CAS Number: 136-53-8
EINECS: 205-251-1
UN: 1268
ADR: 3,III

Order Now

Specification Test Results

Form Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR