Zinc 2-ethylhexanoate (∼20% Zn) ≥97% cont. 1% diethylene glycolmonomethyl ether
Supplier: Thermo Scientific Chemicals
|
Warning
|
39560-22
39560-36
CAAA39560-22EA
85.48
CAD
CAAA39560-22
CAAA39560-36
Zinc 2-ethylhexanoate (∼20% Zn) ≥97% cont. 1% diethylene glycolmonomethyl ether
Zinc 2-ethylhexanoate
CAS No.: 136-53-8
Molecular Formula: C16H30O4Zn
Formula Weight: 351.79
Physical Form: Liquid
MDL No.: MFCD00053316
Formula:
C₁₆H₃₀O₄Zn Storage Temperature: Ambient |
MDL Number:
MFCD00053316 CAS Number: 136-53-8 EINECS: 205-251-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...