Zinc hexafluoro-2,4-pentanedionate dihydrate
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Zinc hexafluoroacetylacetonate hydrate
39559-03
39559-06
CAAA39559-03EA
156.1
CAD
CAAA39559-03
CAAA39559-06
Zinc hexafluoro-2,4-pentanedionate dihydrate
Zinc hexafluoro-2,4-pentanedionate dihydrate
CAS No.: 16743-33-2
Molecular Formula: C10H2F12O4Zn.2H2O
Formula Weight: 515.49 (479.47anhy)
Physical Form: Powder
Melting Point: 157-158°
MDL No.: MFCD00150577
Formula:
C₁₀H₂F₁₂O₄Zn.2H₂O MW: 515.541 g/mol Melting Pt: 157…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD00150577 CAS Number: 16743-33-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...