Zinc naphthenate 67% (10% Zn) in mineral spirits

Supplier: Thermo Scientific Chemicals
Danger

39554-36 39554-A3
CAAA39554-36EA 141.73 CAD
CAAA39554-36 CAAA39554-A3
Zinc naphthenate 67% (10% Zn) in mineral spirits
Zinc naphthenate

CAS No.: 12001-85-3 Flash Point: 37.8° (100°F) Physical Form: Liquid Density: 1.01 Appearance: Colorless UN No.: 1993 MDL No.: MFCD00147699

Formula: 2(C₁₁H₇O₂)·Zn
Density: 1.01
Flash Pt: >37.8° (100 °F)
Storage Temperature: Ambient
MDL Number: MFCD00147699
CAS Number: 12001-85-3
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR