Zinc meso-tetraphenylporphine
Supplier: Thermo Scientific Chemicals
39552-02
39552-04
CAAA39552-02EA
165.9
CAD
CAAA39552-02
CAAA39552-04
Zinc meso-tetraphenylporphine
Zinc meso-tetraphenylporphine
CAS No.: 14074-80-7
Molecular Formula: C44H28N4Zn
Formula Weight: 678.10
Physical Form: Crystalline
Appearance: Purple
Odor: Odorless
MDL No.: MFCD00012155
Formula:
C₄₄H₂₈N₄Zn Storage Temperature: Ambient |
MDL Number:
MFCD00012155 CAS Number: 14074-80-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...