(Pentamethylcyclopentadienyl)zirconium trichloride 99%

Supplier: Thermo Scientific Chemicals
Warning

39551-03 39551-06
CAAA39551-03EA 172.22 CAD
CAAA39551-03 CAAA39551-06
(Pentamethylcyclopentadienyl)zirconium trichloride 99%
(Pentamethylcyclopentadienyl)zirconium trichloride

Purity: 99% CAS No.: 75181-07-6 Molecular Formula: C10H15Cl3Zr Formula Weight: 332.81 Physical Form: Powder. Packaged in ampoules Sensitivity: Moisture Sensitive Appearance: Yellow to yellow-green Melting Point: 230° dec. MDL No.: MFCD00070444

Formula: C₁₀H₁₅Cl₃Zr
MW: 332.81 g/mol
Melting Pt: 230 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00070444
CAS Number: 75181-07-6
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR