Triphenylphosphine-3,3',3''-trisulfonic acid trisodium salt hydrate ≥85%, tech.

Supplier: Thermo Scientific Chemicals
Warning

39538-01 39538-03
CAAAA39538-01EA 240.16 CAD
CAAAA39538-01 CAAA39538-03
Triphenylphosphine-3,3',3''-trisulfonic acid trisodium salt hydrate ≥85%, tech.
Triphenylphosphine-3,3',3''-trisulfonic acid trisodium salt hydrate

Applications: purification of precious metals
Soluble in water


Purity: 85% CAS No.: 63995-70-0 Molecular Formula: C18H12Na3O9PS3.xH2O Formula Weight: 568.41(anhy) Physical Form: Crystalline Solubility: Soluble in water Beilstein Reference: 3584807 MDL No.: MFCD00145472


Applications: In the purification of precious metals

Formula: C₁₈H₁₂Na₃O₉PS₃
Storage Temperature: Ambient
MDL Number: MFCD00145472
CAS Number: 63995-70-0
EINECS: 264-596-6

Order Now

Specification Test Results

Form Crystalline


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR