Palladium hydroxide on carbon ≥99% (Pearlman's catalyst)

Supplier: Thermo Scientific Chemicals

39496-01 39496-03 39496-06
CAAA39496-01EA 139.04 CAD
CAAA39496-01 CAAA39496-03 CAAA39496-06
Palladium hydroxide on carbon ≥99% (Pearlman's catalyst)
Palladium hydroxide

Purity: 99% CAS No.: 12135-22-7 Molecular Formula: Pd(OH)2 Formula Weight: 140.42 Physical Form: Powder Appearance: Black Odor: Odorless MDL No.: MFCD00064599

Formula: H₂O₂Pd
Storage Temperature: Ambient
MDL Number: MFCD00064599
CAS Number: 12135-22-7
EINECS: 235-219-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR