Zirconium titanium oxide ≥99.5% (max. 75ppm Hf)
Supplier: Thermo Scientific Chemicals
39490-14
39490-22
CAAA39490-14EA
151.68
CAD
CAAA39490-14
CAAA39490-22
Zirconium titanium oxide ≥99.5% (max. 75ppm Hf)
Zirconium titanium oxide
Purity: 99.5%
CAS No.: 12036-70-3
Molecular Formula: ZrTiO4
Formula Weight: 203.11
Physical Form: -325 Mesh Powder
Odor: Odorless
MDL No.: MFCD00210648
Formula:
ZrTiO₄ MW: 203.11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00210648 CAS Number: 12036-70-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...