Iron yttrium oxide ≥99.9%
Supplier: Thermo Scientific Chemicals
|
Warning
|
39488-14
39488-22
CAAA39488-14EA
135.41
CAD
CAAA39488-14
CAAA39488-22
Iron yttrium oxide ≥99.9%
Iron yttrium oxide
Purity: 99.9%
CAS No.: 12063-56-8
Molecular Formula: Y3Fe5O12
Formula Weight: 737.94
Physical Form: Powder
Odor: Odorless
MDL No.: MFCD00210618
Formula:
Y₃Fe₅O₁₂ MW: 737.94 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00210618 CAS Number: 12063-56-8 EINECS: 235-053-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...