Nickel(II) cyanide tetrahydrate

Supplier: Thermo Scientific Chemicals
Danger

39480-09 39480-18
CAAA39480-09EA 308.1 CAD
CAAA39480-09 CAAA39480-18
Nickel(II) cyanide tetrahydrate
Nickel(II) cyanide tetrahydrate

Uses: Metallurgy, nickel-plating Insoluble in water. Soluble in ammonium carbonate, ammonia, alkali cyanides


Applications: Metallurgy, nickel-plating
Insoluble in water. Soluble in ammonium carbonate, ammonia, alkali cyanides


Applications: Metallurgy, nickel-plating

Formula: Ni(CN)₂.4H₂O
MW: 182.73 g/mol (110.73 anhy)
Melting Pt: 200 °C (-4H₂O)
Storage Temperature: Ambient
MDL Number: MFCD00049490
CAS Number: 13477-95-7
EINECS: 209-160-8
UN: 1653
ADR: 6.1 II
Merck Index: 14,6506

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR