Nickel(II) octanoate (8% Ni) in mineral spirits

Supplier: Thermo Scientific Chemicals
Danger

39479-14 39479-22
CAAA39479-14EA 86.43 CAD
CAAA39479-14 CAAA39479-22
Nickel(II) octanoate (8% Ni) in mineral spirits
Nickel(II) octanoate

Molecular Formula: C16H30NiO4 Formula Weight: 345.10 Flash Point: 40°(104°F) Physical Form: Liquid Density: 0.95 Appearance: Green UN No.: 1993 MDL No.: MFCD00064239

Formula: C₁₆H₃₀NiO₄
Density: 0.95
Flash Pt: 40 °C (104 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064239
CAS Number: 4995-91-9
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR