Pentaniobium trisilicide ≥99.5% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Niobium silicide
39471-14
39471-22
CAAA39471-14EA
130.19
CAD
CAAA39471-14
CAAA39471-22
Pentaniobium trisilicide ≥99.5% (metals basis)
Pentaniobium trisilicide
Purity: 99.5%
CAS No.: 12060-34-3
Molecular Formula: Nb5Si3
Formula Weight: 548.79
Physical Form: -325 Mesh Powder
Formula:
Nb₅Si₃ MW: 548.79 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00213811 CAS Number: 12060-34-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...