Nickel(II) oxalate dihydrate

Supplier: Thermo Scientific Chemicals
Danger

39454-18 39454-30
CAAA39454-18EA 211.72 CAD
CAAA39454-18 CAAA39454-30
Nickel(II) oxalate dihydrate
Nickel(II) oxalate dihydrate

Soluble in acids. Insoluble in water


CAS No.: 6018-94-6 Molecular Formula: NiC2O4.2H2O Formula Weight: 182.76 (146.73anhy) Physical Form: Powder Odor: Odorless Solubility: Soluble in acids. Insoluble in water UN No.: 3077 MDL No.: MFCD00167155

Formula: C₂H₄NiO₆
Storage Temperature: Ambient
MDL Number: MFCD00167155
CAS Number: 6018-94-6
EINECS: 208-933-7
UN: 3077
ADR: 9,III
Merck Index: 14,6514

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR