Nickel tin oxide dihydrate

Supplier: Thermo Scientific Chemicals
Danger

39452-22 39452-36
CAAA39452-22EA 173.8 CAD
CAAA39452-22 CAAA39452-36
Nickel tin oxide dihydrate
Nickel tin oxide dihydrate

CAS No.: 12035-38-0 Molecular Formula: NiSnO3.2H2O Formula Weight: 261.43 (225.40anhy) Physical Form: Powder Appearance: Pale green Odor: Odorless MDL No.: MFCD00054011

Formula: NiSnO₃.2H₂O
Storage Temperature: Ambient
MDL Number: MFCD00054011
CAS Number: 12035-38-0
EINECS: 234-824-9

Order Now

Specification Test Results

Form Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR