Tetrakis(acetonitrile)palladium(II) tetrafluoroborate (23,5% Pd)
Supplier: Thermo Scientific Chemicals
|
Warning
|
39446-01
39446-03
CAAA39446-01EA
75.05
CAD
CAAA39446-01
CAAA39446-03
Tetrakis(acetonitrile)palladium(II) tetrafluoroborate (23,5% Pd)
Tetrakis(acetonitrile)palladium(II) tetrafluoroborate
Soluble in water
CAS No.: 21797-13-7
Molecular Formula: [Pd(CH3CN)4](BF4)2
Formula Weight: 444.22
Physical Form: Powder
Sensitivity: Air Sensitive
Melting Point: 230° dec.
Solubility: Soluble in water
UN No.: 2811
MDL No.: MFCD00043297
Formula:
C₈H₁₂B₂F₈N₄Pd Melting Pt: 230 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00043297 CAS Number: 21797-13-7 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...